R&D Systems
Manufacturer Code:AF605NA
Catalog # AF605NA
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human Caspase-3 Met1-His277 Accession # AAA65015 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Apopain; Apopain apoptosis-related cysteine protease CASP3 CASP-3 caspase 3 apoptosis-related cysteine peptidase caspase-3 Caspase3 CPP32 CPP-32 CPP32B CPP32SREBP cleavage activity 1 Cysteine protease CPP32 EC 3.4.22 EC 3.4.22.56 LICE-1 PARP cleavage protease procaspase3 Protein Yama SCA-1 Yama; CASP-3; caspase 3, apoptosis-related cysteine peptidase; caspase 3, apoptosis-related cysteine protease; Caspase-3; Caspase-3 subunit p12; Caspase-3 subunit p17; CPP-32; Cysteine protease CPP32; PARP cleavage protease; procaspase3; Protein Yama; SCA-1; SREBP cleavage activity 1
Gene Aliases: CASP3; CPP32; CPP32B; SCA-1
UniProt ID: (Human) P42574
Entrez Gene ID: (Human) 836
Molecular Function: cysteine protease enzyme modulator hydrolase protease protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.