Proteintech
Manufacturer Code:247771AP150UL
Catalog # 50-173-4533
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Caspase 2/p18 Fusion Protein Ag20141 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CASP 2 CASP2 Caspase 2 Caspase 2/p18 Caspase2 ICH 1L ICH 1L/1S ICH1 NEDD 2 NEDD2 Protease ICH 1; CASP-2; caspase 2 apoptosis-related cysteine peptidase; Caspase-2; Caspase-2 subunit p12; Caspase-2 subunit p13; Caspase-2 subunit p18; NEDD-2; Neural precursor cell expressed developmentally down-regulated protein 2; Protease ICH-1; protein phosphatase 1, regulatory subunit 57
Gene Aliases: CASP-2; CASP2; ICH-1; ICH1; NEDD-2; NEDD2; PPP1R57
UniProt ID: (Human) P42575, (Mouse) P29594
Entrez Gene ID: (Human) 835, (Mouse) 12366
Molecular Function:
cysteine protease
enzyme modulator
hydrolase
protease
protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.