Novus Biologicals
Manufacturer Code:NBP290006100UL
Catalog # NB021475
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 101 |
Immunogen | Recombinant Human Caspase-14 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: apoptosis-related cysteine protease; apoptosis-related cysteine protease CASP-14 caspase 14 apoptosis-related cysteine peptidase caspase 14 apoptosis-related cysteine protease caspase-14 EC 3.4.22.- MGC119078 MGC119079 MICE; CASP-14; caspase 14, apoptosis-related cysteine peptidase; caspase 14, apoptosis-related cysteine protease; Caspase-14; Caspase-14 subunit p10, mature form; Caspase-14 subunit p17, mature form; Caspase-14 subunit p20, intermediate form; Caspase-14 subunit p8, intermediate form
Gene Aliases: CASP14
UniProt ID: (Human) P31944
Entrez Gene ID: (Human) 23581
Molecular Function: cysteine protease enzyme modulator hydrolase protease protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.