Novus Biologicals
Manufacturer Code:NBP224518
Catalog # NBP224518
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Within the range of amino acids 100-131 of mouse Caspase-12 was used as the immunogen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Apoptosis Related Cysteine Protease CASP 12 casp12 CASP12P1 Caspase 12 pseudogene 1 Caspase12 UNQ9415; CASP-12; caspase 12 pseudogene 1; Inactive caspase-12
Gene Aliases: CASP-12; CASP12; CASP12P1; UNQ9415/PRO34398
UniProt ID: (Human) Q6UXS9
Entrez Gene ID: (Human) 100506742
Molecular Function: cysteine protease enzyme modulator hydrolase protease protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.