Novus Biologicals
Manufacturer Code:NBP215712
Catalog # NBP215712
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human Caspase 1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: CASP-1; CASP-1 caspase 1 apoptosis-related cysteine peptidase (interleukin 1 beta caspase 1 apoptosis-related cysteine protease (interleukin 1 beta caspase-1 EC 3.4.22 EC 3.4.22.36 ICEP45 IL-1 beta-converting enzyme IL-1BC IL1BC CASP1 nirs variant 1 IL1BCE IL1B-convertase interleukin 1-B converting enzyme interleukin 1-beta convertase Interleukin-1 beta convertase Interleukin-1 beta-converting enzyme p45; CASP1 nirs variant 1; caspase 1, apoptosis-related cysteine peptidase (interleukin 1, beta, convertase); Caspase-1; Caspase-1 subunit p10; Caspase-1 subunit p20; ICE; IL-1 beta-converting enzyme; IL-1BC; IL1B-convertase; interleukin 1, beta, convertase; interleukin 1-B converting enzyme; Interleukin-1 beta convertase; Interleukin-1 beta-converting enzyme; p45
Gene Aliases: CASP1; ICE; IL1BC; IL1BCE; P45
UniProt ID: (Human) P29466
Entrez Gene ID: (Human) 834
Molecular Function:
cysteine protease
enzyme modulator
hydrolase
protease
protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.