Abnova
Manufacturer Code:MAB19412
Catalog # 89-338-916
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | PT55 |
Immunogen | Recombinant protein corresponding to amino acids 318-415 of rat Cask |
Conjugate | ATTO 633 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C |
Mouse Monoclonal Antibody.
Protein Aliases: Calcium/calmodulin-dependent serine protein kinase; Peripheral plasma membrane protein CASK
Gene Aliases: Cask
UniProt ID: (Rat) Q62915
Entrez Gene ID: (Rat) 29647
Molecular Function:
cell junction protein
kinase
nucleotide kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.