Proteintech
Manufacturer Code:224181AP150UL
Catalog # 50-173-3862
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Casein Kinase 2 beta Fusion Protein Ag18134 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alternative name: G5a, phosvitin; casein kinase 2, beta polypeptide; casein kinase II b subunit splicing isoform 132; casein kinase II b subunit splicing isoform 247; casein kinase II b subunit splicing isoform 318; casein kinase II b subunit splicing isoform 660; casein kinase II b subunit splicing isoform 806; Casein kinase II beta subunit; Casein kinase II subunit beta; Casein kinase II subunit beta CK II beta CK2B CK2N CSK2B CSNK2B G5A Phosvitin Protein G5a; chimera CSNK2B-LY6G5B splicing isoform 1072; chimera CSNK2B-LY6G5B splicing isoform 1103; chimera CSNK2B-LY6G5B splicing isoform 532; chimera CSNK2B-LY6G5B splicing isoform 560; chimera CSNK2B-LY6G5B splicing isoform 562; CK II beta; CSNK2B-LY6G5B-1072; CSNK2B-LY6G5B-1103; CSNK2B-LY6G5B-532; CSNK2B-LY6G5B-560; CSNK2B-LY6G5B-562; Phosvitin; Protein G5a
Gene Aliases: CK II beta; CK2B; CK2N; CSK2B; CSNK2B; G5A
UniProt ID: (Human) P67870, (Mouse) P13862
Entrez Gene ID: (Human) 1460, (Mouse) 13001
Molecular Function: enzyme modulator kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.