Novus Biologicals
Manufacturer Code:NBP155002
Catalog # NBP155002
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to CSNK2A1(casein kinase 2 alpha 1 polypeptide) The peptide sequence was selected from the middle region of CSNK2A1. Peptide sequence LGCMLASMIFRKEPFFHGHDNYDQLVRIAKVLGTEDLYDYIDKYNIELDP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: casein kinase 2 alpha 1 polypeptide casein kinase II alpha 1 subunit CK II alpha CK2 catalytic subunit alpha CK2A1casein kinase II subunit alpha CKII EC 2.7.11 EC 2.7.11.1 protein kinase CK2; casein kinase 2, alpha 1 polypeptide; casein kinase II alpha 1 polypeptide pseudogene; casein kinase II alpha 1 subunit; Casein kinase II subunit alpha; CK II alpha; CK II alpha 3; CK2 catalytic subunit alpha; protein kinase CK2
Gene Aliases: CK2A1; CKII; CSNK2A1; CSNK2A3
UniProt ID: (Human) P68400
Entrez Gene ID: (Human) 1457
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.