Proteintech
Manufacturer Code:112301AP150UL
Catalog # 50-172-6986
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Casein Kinase 1 epsilon Fusion Protein Ag1742 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Casein Kinase 1 epsilon casein kinase 1 epsilon CKI epsilon CKIe CSNK1E HCKIE; casein kinase 1, epsilon; Casein kinase I isoform epsilon; CKI, epsilon; CKI-epsilon; CKIe; KC1 epsilon
Gene Aliases: AI426939; AI551861; AW457082; CK1epsilon; CKIe; CKIepsilon; CSNK1E; HCKIE; KC1epsilon; tau
UniProt ID: (Human) P49674, (Mouse) Q9JMK2
Entrez Gene ID: (Human) 1454, (Mouse) 27373, (Rat) 58822
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.