R&D Systems
Manufacturer Code:AF4568
Catalog # AF4568
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human CK1 delta Arg316-Arg415 Accession # P48730 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: casein kinase 1 delta casein kinase I isoform delta CKId CKI-delta CSNK1D EC 2.7.11 EC 2.7.11.1 HCKID; casein kinase 1, delta; Casein kinase I isoform delta; CKI-delta; CKId; Tau-protein kinase CSNK1D
Gene Aliases: ASPS; CKIdelta; CSNK1D; FASPS2; HCKID
UniProt ID: (Human) P48730
Entrez Gene ID: (Human) 1453
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.