Novus Biologicals
Manufacturer Code:NB200341
Catalog # NB200341
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide that maps to a region between residues 450 and 500 of human Coactivator-Associated Arginine Methyltransferase 1 using the numbering given in Swiss-Prot entry Q86X55 (GeneID 10498). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Coactivator-associated arginine methyltransferase 1; coactivator-associated arginine methyltransferase 1EC 2.1.1.- EC 2.1.1 histone-arginine methyltransferase CARM1 PRMT4EC 2.1.1.125 Protein arginine N-methyltransferase 4; Histone-arginine methyltransferase CARM1; Protein arginine N-methyltransferase 4
Gene Aliases: CARM1; PRMT4
UniProt ID: (Human) Q86X55
Entrez Gene ID: (Human) 10498
Molecular Function:
methyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.