R&D Systems
Manufacturer Code:MAB2602100
Catalog # MAB2602100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 89221 |
Immunogen | <em>E. coli</em>-derived recombinant human Cardiotrophin-1 Ser2-Ala201 Accession # Q16619 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: cardiophin 1; cardiophin 1 cardiotrophin 1 cardiotrophin-1 Cardiotrophin1 CT-1CT1 CTF1; Cardiotrophin-1; CT-1
Gene Aliases: CT-1; CT1; CTF1
UniProt ID: (Human) Q16619
Entrez Gene ID: (Human) 1489
Molecular Function:
cytokine
interleukin superfamily
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.