R&D Systems
Manufacturer Code:MAB18741SP
Catalog # MAB18741SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 990007 |
Immunogen | Human Cardiac Troponin T antigen Accession # P45379 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
Protein Aliases: Cardiac muscle troponin T; Cardiac Muscle Troponin T Cardiomyopathy Dilated 1D (Autosomal Dominant) Cardiomyopathy Hypertrophic CMD1D CMH2 CMPD2 cTnT LVNC6 RCM3 TNNT2 TnTC troponin T type 2 (cardiac) troponin T cardiac muscle Troponin T2 Cardiac Type Truncated Cardiac Troponin T; cardiomyopathy, dilated 1D (autosomal dominant); cardiomyopathy, hypertrophic 2; cTnT; TnTc; troponin T type 2 (cardiac); Troponin T, cardiac muscle; troponin T2, cardiac; truncated cardiac troponin T
Gene Aliases: CMD1D; CMH2; CMPD2; cTnT; LVNC6; RCM3; TNNT2; TnTC
UniProt ID: (Human) P45379
Entrez Gene ID: (Human) 7139
Molecular Function: actin binding motor protein actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.