Invitrogen
Manufacturer Code:MA120123
Catalog # PIMA120123
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MF4 |
Immunogen | Free human cardiac troponin and/or native troponin complex. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Cardiac troponin I; cardiac troponin I isoform; cardiomyopathy, dilated 2A (autosomal recessive); CMD2A CMH7 MGC116817 RCM1 TNNC1 cTnI; cTnI; troponin 1, type 3; troponin I type 3 (cardiac); Troponin I, cardiac muscle; troponin I, cardiac muscle-like
Gene Aliases: CMD1FF; CMD2A; CMH7; Ctni; RCM1; TnI; TNNC1; TNNI3
UniProt ID: (Cat) Q863B6, (Human) P19429, (Dog) Q8MKD5, (Bovine) P08057, (Rat) P23693
Entrez Gene ID: (Goat) 100860775, (Cat) 493744, (Human) 7137, (Pig) 100049696, (Dog) 403566, (Bovine) 511094, (Rat) 29248
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.