Invitrogen
Manufacturer Code:MA191566
Catalog # PIMA191566
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 415 |
Immunogen | Synthetic peptide (Human). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Cardiac troponin I; cardiomyopathy, dilated 2A (autosomal recessive); CMD2A CMH7 MGC116817 RCM1 TNNC1 cTnI; troponin I type 3 (cardiac); Troponin I, cardiac muscle
Gene Aliases: CMD1FF; CMD2A; CMH7; cTnI; RCM1; TNNC1; TNNI3
UniProt ID: (Human) P19429
Entrez Gene ID: (Human) 7137
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.