Proteintech
Manufacturer Code:663761IG150UL
Catalog # 50-114-2713
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1D5D6 |
Immunogen | Cardiac Troponin I Fusion Protein Ag16758 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Cardiac troponin I; Cardiac troponin I CMD2A CMH7 cTnI RCM1 TNNC1 TNNI3 troponin I type 3 (cardiac) Troponin I cardiac muscle; cardiomyopathy, dilated 2A (autosomal recessive); troponin 1, type 3; troponin I type 3 (cardiac); Troponin I, cardiac muscle
Gene Aliases: CMD1FF; CMD2A; CMH7; Ctni; RCM1; Tn1; TnI; TNNC1; TNNI3
UniProt ID: (Human) P19429, (Rat) P23693, (Mouse) P48787
Entrez Gene ID: (Human) 7137, (Rat) 29248, (Mouse) 21954
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.