Invitrogen
Manufacturer Code:MA122700
Catalog # PIMA122700
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Inhibition Assays (IA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 19C7 |
Immunogen | Native troponin complex |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Cardiac ankyrin repeat kinase; Cardiac troponin I; cardiac troponin I isoform; Cardiac troponin I-interacting kinase; cardiomyopathy, dilated 2A (autosomal recessive); CMD2A CMH7 MGC116817 RCM1 TNNC1 cTnI; cTnI; Serine/threonine-protein kinase TNNI3K; TNNI3-interacting kinase; TNNI3K; tnni3k {ECO:0000250|UniProtKB:Q59H18}; troponin 1, type 3; troponin I type 3 (cardiac); Troponin I, cardiac muscle; troponin I, cardiac muscle-like
Gene Aliases: CARK; CCDD; CMD1FF; CMD2A; CMH7; Ctni; D830019J24Rik; RCM1; Tn1; TnI; TNNC1; TNNI3; TNNI3K
UniProt ID: (Human) P19429, (Bovine) P08057, (Cat) Q863B6, (Dog) Q8MKD5, (Human) Q59H18, (Mouse) P48787, (Rat) P23693, (Rat) Q7TQP6, (Mouse) Q5GIG6
Entrez Gene ID: (Goat) 100860775, (Human) 7137, (Pig) 100049696, (Bovine) 511094, (Cat) 493744, (Dog) 403566, (Human) 51086, (Dog) 100533996, (Rabbit) 100533992, (Bovine) 535940, (Mouse) 21954, (Rat) 29248, (Rat) 295531, (Mouse) 435766
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
kinase
non-receptor serine/threonine protein kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.