Invitrogen
Manufacturer Code:PA580171
Catalog # PIPA580171
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the N-terminus of human cardiac Troponin C. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cardiac troponin C; slow twitch skeletal/cardiac muscle troponin C; TN-C; Troponin C slow skeletal and cardiac muscles TN-C TNNC1 TNNC; troponin C type 1 (slow); troponin C, cardiac/slow skeletal; Troponin C, slow skeletal and cardiac muscles; troponin C1, slow
Gene Aliases: AI874626; CMD1Z; CMH13; cTnC; cTnI; TN-C; TNC; Tncc; TNNC; TNNC1
UniProt ID: (Human) P63316, (Mouse) P19123
Entrez Gene ID: (Human) 7134, (Rat) 290561, (Mouse) 21924
Molecular Function:
actin family cytoskeletal protein
calcium-binding protein
calmodulin
cytoskeletal protein
intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.