Invitrogen
Manufacturer Code:PA576641
Catalog # PIPA576641
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human Cardiac Troponin C. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cardiac muscleCardiac troponin ICardiomyopathy familial hypertrophic 7 includedCMD1FFCMD2ACMH7cTnIFamilial hypertrophic cardiomyopathy 7MGC116817RCM1Tn1TniTNN I3TNNC 1TNNC1TNNI3TNNI3&lowbarHUMANTroponin ITroponin I cardiacTroponin I cardiac muscleTroponin I cardiac muscle isoformTroponin I type 3 cardiactroponin I cardiac 3TroponinITtroponin I type 3 (cardiac); cardiac troponin C; slow twitch skeletal/cardiac muscle troponin C; TN-C; troponin C type 1 (slow); Troponin C, slow skeletal and cardiac muscles; troponin C1, slow
Gene Aliases: AI874626; CMD1Z; CMH13; cTnC; cTnI; TN-C; TNC; Tncc; TNNC; TNNC1
UniProt ID: (Human) P63316, (Mouse) P19123
Entrez Gene ID: (Human) 7134, (Mouse) 21924
Molecular Function: actin family cytoskeletal protein calcium-binding protein calmodulin cytoskeletal protein intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.