Invitrogen
Manufacturer Code:PA5116462
Catalog # PIPA5116462
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant full length protein of human TNNC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI874626 cardiac 3 cardiac troponin C Cardiac troponin I CMD1FF CMD1Z CMD2A CMH13 CMH7 cTnC cTnI MGC116817 RCM1 slow twitch skeletal/cardiac muscle troponin C Tn1 TnC TN-C tncc Tni TNN I3 TNNC TNNC 1 TNNC1 TNNI3 troponin C type 1 (slow) troponin C cardiac/slow skeletal troponin C slow skeletal and cardiac muscles troponin C1 slow troponin C1 slow skeletal and cardiac type Troponin I Troponin I cardiac TroponinI; cardiac troponin C; slow twitch skeletal/cardiac muscle troponin C; TN-C; troponin C type 1 (slow); troponin C, cardiac/slow skeletal; Troponin C, slow skeletal and cardiac muscles; troponin C1, slow
Gene Aliases: AI874626; CMD1Z; CMH13; cTnC; cTnI; TN-C; TNC; Tncc; TNNC; TNNC1
UniProt ID: (Human) P63316, (Mouse) P19123
Entrez Gene ID: (Human) 7134, (Rat) 290561, (Mouse) 21924
Molecular Function:
actin family cytoskeletal protein
calcium-binding protein
calmodulin
cytoskeletal protein
intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.