Novus Biologicals
Manufacturer Code:NBP29265520UL
Catalog # NB027180
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 257-476 of human CPE (NP_001864.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Carboxypeptidase E; carboxypeptidase E Carboxypeptidase H CPH EC 3.4.17 EC 3.4.17.10 Enkephalin convertase Prohormone-processing carboxypeptidase; Carboxypeptidase H; cobalt-stimulated chromaffin granule carboxypeptidase; CPE; CPH; Enkephalin convertase; insulin granule-associated carboxypeptidase; Prohormone-processing carboxypeptidase
Gene Aliases: CPE; CPH
UniProt ID: (Human) P16870
Entrez Gene ID: (Human) 1363
Molecular Function:
hydrolase
metalloprotease
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.