Invitrogen
Manufacturer Code:PA577027
Catalog # PIPA577027
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human Carboxypeptidase E. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Carboxypeptidase E; Carboxypeptidase E Carboxypeptidase H Carboxypeptidase H precursor CarboxypeptidaseE CarboxypeptidaseH CBPE Cobalt stimulated chromaffin granule carboxypeptidase CPE Cph 1 CPH Cph1 Enkephalin convertase Entephalin convertase Insulin granule associated carboxypeptidase Prohormone processing carboxypeptidase Prohormone-processing carboxypeptidase; Carboxypeptidase H; cobalt-stimulated chromaffin granule carboxypeptidase; CPE; CPH; Enkephalin convertase; insulin granule-associated carboxypeptidase; Prohormone-processing carboxypeptidase
Gene Aliases: CPE; CPH; Cph-1; Cph1; fat; R74677
UniProt ID: (Human) P16870, (Mouse) Q00493
Entrez Gene ID: (Human) 1363, (Mouse) 12876
Molecular Function: hydrolase metalloprotease protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.