Invitrogen
Manufacturer Code:PA570395
Catalog # PIPA570395
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N terminal region of human Carboxypeptidase E |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Carboxypeptidase E; Carboxypeptidase H; cobalt-stimulated chromaffin granule carboxypeptidase; CPE; CPH; CPH carboxypeptidase H cobalt-stimulated chromaffin granule carboxypeptidase enkephalin convertase insulin granule-associated carboxypeptidase prohormone-processing carboxypeptidase; Enkephalin convertase; insulin granule-associated carboxypeptidase; Prohormone-processing carboxypeptidase; unnamed protein product
Gene Aliases: CARBE; CPE; CPH; Cph-1; Cph1; fat; R74677; si:ch211-198g9.2; zgc:85981
UniProt ID: (Human) P16870, (Bovine) P04836, (Rat) P15087, (Mouse) Q00493
Entrez Gene ID: (Human) 1363, (Bovine) 280753, (Rabbit) 108175425, (Rat) 25669, (Mouse) 12876, (Guinea pig) 100721385, (Zebrafish) 407979
Molecular Function: hydrolase metalloprotease protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.