Invitrogen
Manufacturer Code:PA5109581
Catalog # PIPA5109581
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 257-476 of human CPE (NP_001864.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BOS_16236 CARBE Carboxypeptidase E Carboxypeptidase H CarboxypeptidaseE CarboxypeptidaseH CBPE Cpe CPH Cph 1 Cph1 Cph-1 Enkephalin convertase fat Prohormone-processing carboxypeptidase R74677 si:ch211-198g9.2 zgc:85981; Carboxypeptidase E; Carboxypeptidase H; cobalt-stimulated chromaffin granule carboxypeptidase; CPE; CPH; Enkephalin convertase; insulin granule-associated carboxypeptidase; Prohormone-processing carboxypeptidase
Gene Aliases: CPE; CPH; Cph-1; Cph1; fat; R74677
UniProt ID: (Human) P16870, (Mouse) Q00493
Entrez Gene ID: (Human) 1363, (Mouse) 12876
Molecular Function:
hydrolase
metalloprotease
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.