Invitrogen
Manufacturer Code:PA131035
Catalog # PIPA131035
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide conjugated to KLH (C terminal). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Carboxypeptidase E; Carboxypeptidase H; cobalt-stimulated chromaffin granule carboxypeptidase; CPE; CPH; Enkephalin convertase; insulin granule-associated carboxypeptidase; Prohormone-processing carboxypeptidase; unnamed protein product
Gene Aliases: CARBE; CPE; CPH; Cph-1; Cph1; fat; R74677
UniProt ID: (Human) P16870, (Bovine) P04836, (Rat) P15087, (Mouse) Q00493
Entrez Gene ID: (Human) 1363, (Bovine) 280753, (Rat) 25669, (Mouse) 12876
Molecular Function:
hydrolase
metalloprotease
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.