Invitrogen
Manufacturer Code:PA575825
Catalog # PIPA575825
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues in Human Carboxypeptidase D. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Carboxypeptidase D; Carboxypeptidase D Metallocarboxypeptidase D gp180 CPD; cpd {ECO:0000312|RGD:2393}; gp180; Metallocarboxypeptidase D
Gene Aliases: AA960140; AW322530; CPD; D830034L15Rik; GP180
UniProt ID: (Human) O75976, (Rat) Q9JHW1, (Mouse) O89001
Entrez Gene ID: (Human) 1362, (Rat) 25306, (Mouse) 12874
Molecular Function: hydrolase metalloprotease protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.