Invitrogen
Manufacturer Code:PA579063
Catalog # PIPA579063
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived human CPB2 recombit protein (Position: K166-D388). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: carboxypeptidase B-like protein; Carboxypeptidase B2; carboxypeptidase B2 (plasma); carboxypeptidase B2 (plasma, carboxypeptidase U); carboxypeptidase R; Carboxypeptidase U; CPU; pCPB; Plasma carboxypeptidase B; TAFI; Thrombin-activable fibrinolysis inhibitor; thrombin-activatable fibrinolysis inhibitor
Gene Aliases: CPB2; CPU; PCPB; TAFI
UniProt ID: (Human) Q96IY4
Entrez Gene ID: (Human) 1361
Molecular Function: hydrolase metalloprotease protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.