Invitrogen
Manufacturer Code:PA569156
Catalog # PIPA569156
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse Carboxypeptidase A3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Carboxypeptidase A3; carboxypeptidase A3 (mast cell); Mast cell carboxypeptidase A; MC-CPA; MC-CPA mast cell carboxypeptidase A tissue carboxypeptidase A; R-CPA; RMC-CP; tissue carboxypeptidase A
Gene Aliases: CPA3; MC-CPA; mMC-CPA
UniProt ID: (Human) P15088, (Rat) P21961, (Mouse) P15089
Entrez Gene ID: (Bovine) 528454, (Human) 1359, (Dog) 485707, (Rat) 54242, (Mouse) 12873, (Guinea pig) 100715728
Molecular Function: hydrolase metalloprotease protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.