Proteintech
Manufacturer Code:162361AP150UL
Catalog # 50-173-0816
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carboxypeptidase A3 Fusion Protein Ag8984 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Carboxypeptidase A3; carboxypeptidase A3 (mast cell); Carboxypeptidase A3 CPA3 Mast cell carboxypeptidase A MC CPA; Mast cell carboxypeptidase A; MC-CPA; R-CPA; RMC-CP; tissue carboxypeptidase A
Gene Aliases: CPA3; MC-CPA; mMC-CPA
UniProt ID: (Human) P15088, (Rat) P21961, (Mouse) P15089
Entrez Gene ID: (Human) 1359, (Rat) 54242, (Mouse) 12873
Molecular Function: hydrolase metalloprotease protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.