Novus Biologicals
Manufacturer Code:NBP270443V
Catalog # NBP270443V
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3D8 |
Immunogen | Full length recombinant protein of human Carboxypeptidase A2/CPA2 (NP_001860) produced in HEK293T cells. |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Carboxypeptidase A2; carboxypeptidase A2 (pancreatic); carboxypeptidase A2 carboxypeptidase A2 (pancreatic) EC 3.4.17 EC 3.4.17.15
Gene Aliases: CPA2
UniProt ID: (Human) P48052
Entrez Gene ID: (Human) 1358
Molecular Function:
hydrolase
metalloprotease
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.