R&D Systems
Manufacturer Code:AF5657SP
Catalog # AF5657SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Carboxylesterase 2/CES2 Gln27-Leu559 Accession # O00748 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Carboxylesterase 2; carboxylesterase 2 (intestine, liver); CE-2; Cocaine esterase; hCE-2; intestinal carboxylesterase; liver carboxylesterase-2; Methylumbelliferyl-acetate deacetylase 2
Gene Aliases: CE-2; CES2; CES2A1; ICE; PCE-2
UniProt ID: (Human) O00748
Entrez Gene ID: (Human) 8824
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.