Novus Biologicals
Manufacturer Code:NBP157922
Catalog # NBP157922
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to CEL(carboxyl ester lipase (bile salt-stimulated lipase)) The peptide sequence was selected from the middle region of CEL. Peptide sequence VTPTGDSETAPVPPTGDSGAPPVPPTGDSEAAPVPPTDDSKEAQMPAVIR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BAL bile salt-activated lipase Bile salt-stimulated lipase BSDL BSSLbile salt-dependent lipase oncofetal isoform bucelipase carboxyl ester hydrolase Carboxyl ester lipase carboxyl ester lipase (bile salt-stimulated lipase) CEase CELL Cholesterol esterase EC 3.1.1 EC 3.1.1.13 EC 3.1.1.3 FAP FAPP fetoacinar pancreatic protein LIPA lysophospholipase pancreatic MODY8bile-salt-activated lipase Pancreatic lysophospholipase Sterol esterase; bile salt-dependent lipase, oncofetal isoform; bucelipase; carboxyl ester hydrolase; carboxyl ester lipase (bile salt-stimulated lipase); cholesterol esterase; fetoacinar pancreatic protein; lysophospholipase, pancreatic; sterol esterase
Gene Aliases: BAL; BSDL; BSSL; CEase; CELL; FAP; FAPP; LIPA; MODY8
Entrez Gene ID: (Human) 1056
Molecular Function:
esterase
hydrolase
lipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.