Novus Biologicals
Manufacturer Code:NBP298314AF594
Catalog # NB053325
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant Carboxyl Ester Lipase/CEL (NP_034015.1 Met1-Leu534). |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BAL bile salt-activated lipase Bile salt-stimulated lipase BSDL BSSLbile salt-dependent lipase oncofetal isoform bucelipase carboxyl ester hydrolase Carboxyl ester lipase carboxyl ester lipase (bile salt-stimulated lipase) CEase CELL Cholesterol esterase EC 3.1.1 EC 3.1.1.13 EC 3.1.1.3 FAP FAPP fetoacinar pancreatic protein LIPA lysophospholipase pancreatic MODY8bile-salt-activated lipase Pancreatic lysophospholipase Sterol esterase; bile salt-dependent lipase, oncofetal isoform; bucelipase; carboxyl ester hydrolase; carboxyl ester lipase (bile salt-stimulated lipase); cholesterol esterase; fetoacinar pancreatic protein; lysophospholipase, pancreatic; sterol esterase
Gene Aliases: BAL; BSDL; BSSL; CEase; CELL; FAP; FAPP; LIPA; MODY8
Entrez Gene ID: (Human) 1056
Molecular Function: esterase hydrolase lipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.