Invitrogen
Manufacturer Code:MA529662
Catalog # PIMA529662
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 1 |
Immunogen | Recombit Mouse Carbonic Anhydrase IX/CAR9 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: CA-IX; CA9 CAIX Carbonic dehydratase Membrane antigen MN P54 / 58N pMW1 RCC associated protein G250 Renal cell carcinoma associated antigen G250; Carbonate dehydratase IX; Carbonic anhydrase 9; Carbonic anhydrase IX; Membrane antigen MN homolog
Gene Aliases: Ca9; CAIX; Car9; MN/CA9
UniProt ID: (Mouse) Q8VHB5
Entrez Gene ID: (Mouse) 230099
Molecular Function:
dehydratase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.