Invitrogen
Manufacturer Code:PA586519
Catalog # PIPA586519
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 50-104 of Human CA IX. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CA-IX; Carbonate dehydratase IX; Carbonic anhydrase 9; Carbonic anhydrase 9 Carbonate dehydratase IX Carbonic anhydrase IX CA-IX CAIX Membrane antigen MN P54/58N Renal cell carcinoma-associated antigen G250 RCC-associated antigen G250 pMW1 CA9 G250 MN; Carbonic anhydrase IX; carbonic dehydratase; Membrane antigen MN; Membrane antigen MN homolog; P54/58N; pMW1; RCC-associated antigen G250; RCC-associated protein G250; Renal cell carcinoma-associated antigen G250
Gene Aliases: CA9; CAIX; Car9; G250; MN; MN/CA9
UniProt ID: (Human) Q16790, (Mouse) Q8VHB5
Entrez Gene ID: (Human) 768, (Mouse) 230099, (Rat) 313495
Molecular Function:
dehydratase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.