Novus Biologicals
Manufacturer Code:NBP28953850UL
Catalog # NB020560
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 6 |
Immunogen | Recombinant Human Carbonic Anhydrase III/CA3 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: CA-III; CAIII CA-III Car3 Carbonate dehydratase III carbonic anhydrase 3 Carbonic anhydrase III carbonic anhydrase III muscle specific EC 4.2.1.1; Carbonate dehydratase III; Carbonic anhydrase 3; Carbonic anhydrase III; carbonic anhydrase III, muscle specific; epididymis secretory sperm binding protein Li 167mP; HEL-S-167mP
Gene Aliases: CA3; CAIII; Car3
UniProt ID: (Human) P07451
Entrez Gene ID: (Human) 761
Molecular Function:
dehydratase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.