Novus Biologicals
Manufacturer Code:NBP289537B
Catalog # NB108208
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 2 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human Carbonic Anhydrase III/CA3 (NP_005172.1 Met1-Lys260). |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: CA-III; CAIII CA-III Car3 Carbonate dehydratase III carbonic anhydrase 3 Carbonic anhydrase III carbonic anhydrase III muscle specific EC 4.2.1.1; Carbonate dehydratase III; Carbonic anhydrase 3; Carbonic anhydrase III; carbonic anhydrase III, muscle specific; epididymis secretory sperm binding protein Li 167mP; HEL-S-167mP
Gene Aliases: CA3; CAIII; Car3
UniProt ID: (Human) P07451
Entrez Gene ID: (Human) 761
Molecular Function: dehydratase lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.