Invitrogen
Manufacturer Code:PA578899
Catalog # PIPA578899
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived human CA3 recombit protein (Position: D14-E235). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CA-III; Carbonate dehydratase III; Carbonic anhydrase 3; Carbonic anhydrase 3 Carbonate dehydratase III Carbonic anhydrase III CA-III CA3; Carbonic anhydrase III; carbonic anhydrase III, muscle specific; epididymis secretory sperm binding protein Li 167mP; HEL-S-167mP
Gene Aliases: BB219044; CA3; CAIII; Car-3; Car3
UniProt ID: (Human) P07451, (Mouse) P16015, (Rat) P14141
Entrez Gene ID: (Human) 761, (Mouse) 12350, (Rat) 54232
Molecular Function: dehydratase lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.