Novus Biologicals
Manufacturer Code:NBP289522G
Catalog # NB108099
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 1 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human Carbonic Anhydrase II/CA2 (NP_000058.1 Met1-Lys260). |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: CA-II; CAC; CAII CA-IIEC 4.2.1.1 Car2 Carbonate dehydratase II carbonic anhydrase 2 carbonic anhydrase B Carbonic anhydrase C carbonic anhydrase IICAC carbonic dehydratase; Carbonate dehydratase II; Carbonic anhydrase 2; carbonic anhydrase B; Carbonic anhydrase C; Carbonic anhydrase II; carbonic dehydratase; epididymis luminal protein 76; epididymis secretory protein Li 282
Gene Aliases: CA-II; CA2; CAC; CAII; Car2; HEL-76; HEL-S-282
UniProt ID: (Human) P00918
Entrez Gene ID: (Human) 760
Molecular Function: dehydratase lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.