Invitrogen
Manufacturer Code:MA538188
Catalog # PIMA538188
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1451 |
Immunogen | A synthesized peptide derived from human Carbonic anhydrase 2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: AI131712 CA II Ca2 CAC CAII CA-II Car2 Car-2 carbonate dehydratase II Carbonic anhydrase 2 carbonic anhydrase B carbonic anhydrase C Carbonic anhydrase II carbonic dehydratase epididymis luminal protein 76 epididymis secretory protein Li 282 HEL-76 HEL-S-282 Ltw-5 Lvtw-5; CA II; CA-II; CAC; Carbonate dehydratase II; Carbonic anhydrase 2; carbonic anhydrase B; Carbonic anhydrase C; Carbonic anhydrase II; carbonic dehydratase; epididymis luminal protein 76; epididymis secretory protein Li 282
Gene Aliases: AI131712; CA-II; CA2; CAC; CAII; Car-2; Car2; HEL-76; HEL-S-282; Ltw-5; Lvtw-5
UniProt ID: (Human) P00918, (Mouse) P00920, (Rat) P27139
Entrez Gene ID: (Human) 760, (Mouse) 12349, (Rat) 54231
Molecular Function: dehydratase lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.