Invitrogen
Manufacturer Code:MA530206
Catalog # PIMA530206
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 2 |
Immunogen | Recombit Mouse Carbonic Anhydrase II/CA2 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: CA II; CA-II; CA-II CAII Car2 CAC carbonate dehydratase II carbonic anhydrase 2 carbonic anhydrase B carbonic anhydrase C carbonic dehydratase; Carbonate dehydratase II; Carbonic anhydrase 2; Carbonic anhydrase II
Gene Aliases: AI131712; Ca2; CAII; Car-2; Car2; Ltw-5; Lvtw-5
UniProt ID: (Mouse) P00920
Entrez Gene ID: (Mouse) 12349
Molecular Function: dehydratase lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.