Invitrogen
Manufacturer Code:PA580391
Catalog # PIPA580391
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human Carbonic Anhydrase II/CA2 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CA-II; CA-II CAII Car2 CAC carbonate dehydratase II carbonic anhydrase 2 carbonic anhydrase B carbonic anhydrase C carbonic dehydratase; CAC; Carbonate dehydratase II; Carbonic anhydrase 2; carbonic anhydrase B; Carbonic anhydrase C; Carbonic anhydrase II; carbonic dehydratase; epididymis luminal protein 76; epididymis secretory protein Li 282
Gene Aliases: CA-II; CA2; CAC; CAII; Car2; HEL-76; HEL-S-282
UniProt ID: (Human) P00918
Entrez Gene ID: (Human) 760
Molecular Function:
dehydratase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.