Novus Biologicals
Manufacturer Code:NBP191748
Catalog # NBP191748
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:TSRMYEFVGLEFLPHLQTWVHNITRGKGMGDHAFHTNARDALNVSQAWRWSLPYEKVSRLQKACGDAMNLLGYRHVRSEQEQRNLLLDLLSTWTVPEQIH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 4; carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 4 EC 2.8.2 EC 2.8.2.- EC 2.8.2.17 EC 2.8.2.21 galactose/N-acetylglucosamine/N-acetylgalactosamine 6-O-sulfotransferase 3 Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 3 glcNAc6ST-2 GlcNAc6ST2 Gn6st-2 GST3 GST-3 HEC-GlcNAc6ST HECGLCNAC6ST HEC-GLCNAC-6-ST High endothelial cells N-acetylglucosamine 6-O-sulfotransferase L-selectin ligand sulfotransferase LSSTcarbohydrate sulfotransferase 4 N-acetylglucosamine 6-O-sulfotransferase 2; Carbohydrate sulfotransferase 4; galactose/N-acetylglucosamine/N-acetylgalactosamine 6-O-sulfotransferase 3; Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 3; GlcNAc6ST-2; gn6st-2; GST-3; HEC-GlcNAc6ST; High endothelial cells N-acetylglucosamine 6-O-sulfotransferase; L-selectin ligand sulfotransferase; LSST; N-acetylglucosamine 6-O-sulfotransferase 2
Gene Aliases: CHST4; GlcNAc6ST2; GST3; HECGLCNAC6ST; LSST
UniProt ID: (Human) Q8NCG5
Entrez Gene ID: (Human) 10164
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.