R&D Systems
Manufacturer Code:FAB36551RR100UG
Catalog # FAB36551RR1
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 352110R |
Immunogen | Human embryonic kidney cell line HEK293-derived transfected with human Cannabinoid R2/CB2/CNR2 Met1-Cys360 Accession # NP_001832 |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8°C as supplied. |
Mouse Monoclonal Antibody
Protein Aliases: Cannabinoid receptor 2; cannabinoid receptor 2 (macrophage); cannabinoid receptor 2 (macrophage) CannabinoidR2 CB-2 CB2cannabinoid receptor 2 CB2R CNR2 CX5 CX5testis-dominant CNR2 isoform CB2 hCB2; CB-2; CX5; testis-dominant CNR2 isoform CB2
Gene Aliases: CB-2; CB2; CB2A; CB2B; CNR2; CX5
UniProt ID: (Human) P34972
Entrez Gene ID: (Human) 1269
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.