R&D Systems
Manufacturer Code:FAB3834A
Catalog # FAB3834A
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 368302 |
Immunogen | HEK293 human embryonic kidney cell line transfected with human Cannabinoid R1/CB1/CNR1 Met1-Leu472 Accession # NP_057167 |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse Monoclonal Antibody
Protein Aliases: CANN6; CANN6 CANN6CB1R cannabinoid receptor 1 (brain) CannabinoidR1 CB1 CB1A CB1K5 CB1R CB-R CB-RCB1cannabinoid receptor 1 central cannabinoid receptor CNR CNR1 SKR6R; Cannabinoid receptor 1; cannabinoid receptor 1 (brain); CB-R; central cannabinoid receptor
Gene Aliases: CANN6; CB-R; CB1; CB1A; CB1K5; CB1R; CNR; CNR1
UniProt ID: (Human) P21554
Entrez Gene ID: (Human) 1268
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.