Abnova
Manufacturer Code:PAB11208
Catalog # 89-114-923
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 457-474 of rat Camk4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Rabbit polyclonal antibody raised against synthetic peptide of Camk4.
For Research Use Only
Protein Aliases: Calcium/calmodulin-dependent protein kinase type IV; Calmodulin-dependent protein kinase IV; Calspermin; CAM kinase-GR; CaMK IV; Ccdpk; RATCCDPK
Gene Aliases: Camk4; Ccdpk; RATCCDPK
UniProt ID: (Rat) P13234
Entrez Gene ID: (Rat) 25050
Molecular Function: cytoskeletal protein kinase microtubule family cytoskeletal protein non-motor microtubule binding protein non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.