Invitrogen
Manufacturer Code:PA585406
Catalog # PIPA585406
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human Calsequestrin 1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Aspartactin; calmitin; Calmitine; calsequestrin 1 (fast-twitch, skeletal muscle); Calsequestrin, skeletal muscle isoform; Calsequestrin-1; CASQ antibody PDIB1 antibody CASQ1 antibody calmitine antibody calsequestrin skeletal muscle isoform antibody calmitin antibody calsequestrin-1 antibody calsequestrin 1 (fast-twitch skeletal muscle) antibody; Laminin-binding protein
Gene Aliases: CASQ; CASQ1; CSQ; CSQ-1; CSQ1; PDIB1; sCSQ; VMCQA
UniProt ID: (Human) P31415, (Rat) P19633, (Mouse) O09165
Entrez Gene ID: (Human) 844, (Rat) 686019, (Mouse) 12372
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.