Proteintech
Manufacturer Code:275601AP150UL
Catalog # 50-173-5701
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Calsequestrin 2 Fusion Protein Ag26412 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: calsequestrin 2 (cardiac muscle); Calsequestrin 2 CASQ2 PDIB2; calsequestrin 2, fast-twitch, cardiac muscle; Calsequestrin, cardiac muscle isoform; Calsequestrin-2; cardiac calsequestrin
Gene Aliases: AA033488; AW146219; CASQ2; cCSQ; Csq2; ESTM52; PDIB2
UniProt ID: (Human) O14958, (Mouse) O09161
Entrez Gene ID: (Human) 845, (Mouse) 12373
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.