Novus Biologicals
Manufacturer Code:NBP147518
Catalog # NBP147518
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
In vitro Assay (IV) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1G6A7 |
Immunogen | A synthetic peptide corresponding to the C-terminus (EEEDVPGQAKDELC) of human Calreticulin conjugated to KLH. [UniProt# P27797] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: autoantigen Ro calregulin calreticulin cC1qR CRP55 CRT CRTC Endoplasmic reticulum resident protein 60 ERp60 FLJ26680 grp60 RO Sicca syndrome antigen A (autoantigen Ro calreticulin) SSAHACBP; Calregulin; Calreticulin; CRP55; Endoplasmic reticulum resident protein 60; epididymis secretory sperm binding protein Li 99n; ERp60; grp60; HACBP; Sicca syndrome antigen A (autoantigen Ro; calreticulin)
Gene Aliases: CALR; cC1qR; CRT; CRTC; HEL-S-99n; RO; SSA
UniProt ID: (Human) P27797
Entrez Gene ID: (Human) 811
Molecular Function:
calcium-binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.