Proteintech
Manufacturer Code:248551AP150UL
Catalog # 50-173-4578
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Calponin Fusion Protein Ag21384 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Basic calponin; Basic calponin Calponin Calponin 1 Calponin H1 smooth muscle CNN1 Sm Calp SMCC; calponin 1, basic, smooth muscle; Calponin H1, smooth muscle; Calponin-1; calponins, basic; epididymis secretory protein Li 14
Gene Aliases: CN; CNN1; CnnI; HEL-S-14; Sm-Calp; SMCC
UniProt ID: (Human) P51911, (Mouse) Q08091
Entrez Gene ID: (Human) 1264, (Mouse) 12797
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.